siRNA Details
| Gene Symbol: Frmpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081172.2 | NP_001074641.1 | 16 | 4616 | ||
| GenBank mRNA | AK122409.1 | - | 13 | 4242 | ||
| AK133351.1 | - | 3 | 3055 | |||
| BC031840.1 | - | 6 | 3476 | |||
| BC046469.1 | - | 2 | 2392 | |||
| BC051097.1 | - | 6 | 3476 | |||
| BC158060.1 | - | 16 | 4580 | 9007360 | ||
| BG070589.2 | - | 1 | 193 | |||

Frmpd1
Genomic Map