siRNA Details
| Gene Symbol: Frmpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081172.2 | NP_001074641.1 | 11 | 1130 | ||
| GenBank mRNA | AK122409.1 | - | 8 | 756 | ||
| BC158060.1 | - | 11 | 1094 | 9007360 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmpd1| Gene Symbol: Frmpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081172.2 | NP_001074641.1 | 11 | 1130 | ||
| GenBank mRNA | AK122409.1 | - | 8 | 756 | ||
| BC158060.1 | - | 11 | 1094 | 9007360 | ||