siRNA Details
| Gene Symbol: Zfp462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172867.3 | NP_766455.2 | 9 | 7080 | ||
| GenBank mRNA | AK031378.1 | - | 4 | 591 | ||
| AK173266.1 | - | 7 | 1393 | |||
| BC099692.1 | - | 4 | 677 | |||
| DQ355518.1 | - | 8 | 6777 | |||

Zfp462
Genomic Map