siRNA Details
| Gene Symbol: Rasef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017427.1 | NP_001017427.1 | 5, 6 | 652 | ||
| GenBank mRNA | AK132691.1 | - | 5, 6 | 632 | ||
| BC030882.1 | - | 1, 2 | 49 | |||
| BC040766.1 | - | 1, 2 | 38 | |||
| BC125539.1 | - | 4, 5 | 466 | 40130054 | ||
| BC137774.1 | - | 4, 5 | 466 | 8860793 | ||
| BC144901.1 | - | 4, 5 | 466 | |||

Rasef
Genomic Map