siRNA Details
| Gene Symbol: Wdr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146254.4 | NP_666366.4 | 4 | 574 | ||
| GenBank mRNA | BC028975.1 | - | 4 | 499 | 5363117 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr78| Gene Symbol: Wdr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146254.4 | NP_666366.4 | 4 | 574 | ||
| GenBank mRNA | BC028975.1 | - | 4 | 499 | 5363117 | |