siRNA Details
| Gene Symbol: Zyg11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033634.3 | NP_001028806.2 | 5 | 1327 | ||
| GenBank mRNA | AK145130.1 | - | 5 | 1300 | ||
| AK148335.1 | - | 5 | 1326 | |||
| AK162646.1 | - | 5 | 1290 | |||
| AK166031.1 | - | 5 | 1307 | |||
| AK166174.1 | - | 5 | 1307 | |||
| AK166278.1 | - | 5 | 1317 | |||
| BC117977.2 | - | 5 | 1291 | 40099428 | ||
| BC117978.2 | - | 5 | 1291 | 40099429 | ||
| BC139383.1 | - | 5 | 1294 | 8862405 | ||
| BC139384.1 | - | 5 | 1294 | 8862406 | ||

Zyg11b
Genomic Map