siRNA Details
| Gene Symbol: Zyg11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033634.3 | NP_001028806.2 | 2 | 342 | ||
| GenBank mRNA | AK145130.1 | - | 2 | 315 | ||
| AK148335.1 | - | 2 | 341 | |||
| AK162646.1 | - | 2 | 305 | |||
| AK166031.1 | - | 2 | 322 | |||
| AK166174.1 | - | 2 | 322 | |||
| AK166278.1 | - | 2 | 332 | |||
| BC117977.2 | - | 2 | 306 | 40099428 | ||
| BC117978.2 | - | 2 | 306 | 40099429 | ||
| BC139383.1 | - | 2 | 309 | 8862405 | ||
| BC139384.1 | - | 2 | 309 | 8862406 | ||

Zyg11b
Genomic Map