siRNA Details
| Gene Symbol: Ifnlr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174851.3 | NP_777276.3 | 7 | 1303 | ||
| GenBank mRNA | AK141338.1 | - | 7 | 1304 | ||
| AK205737.1 | - | 1 | 277 | |||
| AY184376.1 | - | 7 | 1218 | |||
| BC057856.1 | - | 7 | 1236 | 5036481 | ||

Ifnlr1
Genomic Map