siRNA Details
| Gene Symbol: Slc45a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173774.3 | NP_776135.2 | 5 | 1230 | ||
| GenBank mRNA | AK082651.1 | - | 1223 | |||
| AK134677.1 | - | 5 | 1156 | |||
| AK134683.1 | - | 5 | 1220 | |||
| AK158002.1 | - | 5 | 1220 | |||
| BC051188.1 | - | 5 | 1192 | |||

Slc45a1
Genomic Map