siRNA Details
| Gene Symbol: Napepld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178728.5 | NP_848843.1 | 3 | 693 | ||
| GenBank mRNA | AB112350.1 | - | 2 | 536 | ||
| AK041194.1 | - | 3 | 691 | |||
| AK049135.1 | - | 3 | 952 | |||
| AK080888.1 | - | 3 | 693 | |||
| AK141585.1 | - | 3 | 973 | |||
| BC062890.1 | - | 3 | 715 | 5702354 | ||

Napepld
Genomic Map