siRNA Details
Gene Symbol: Cpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153107.2 | NP_694747.2 | 3 | 352 | ||
GenBank mRNA | AF356844.1 | - | 3 | 261 | ||
CJ067998.1 | - | 3 | 424 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153107.2 | NP_694747.2 | 3 | 352 | ||
GenBank mRNA | AF356844.1 | - | 3 | 261 | ||
CJ067998.1 | - | 3 | 424 |