siRNA Details
| Gene Symbol: Tmprss11f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178730.3 | NP_848845.1 | 11 | 1443 | ||
| GenBank mRNA | AK028447.1 | - | 11 | 1445 | ||
| AK028587.1 | - | 11 | 1438 | |||
| AK029032.1 | - | 11 | 1405 | |||
| AK037173.1 | - | 10 | 1287 | |||
| BC132629.1 | - | 10 | 1293 | 40130906 | ||

Tmprss11f
Genomic Map