siRNA Details
| Gene Symbol: Tmprss11f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178730.3 | NP_848845.1 | 8 | 783 | ||
| GenBank mRNA | AK028447.1 | - | 8 | 785 | ||
| AK028587.1 | - | 8 | 778 | |||
| AK029032.1 | - | 8 | 745 | |||
| AK037173.1 | - | 7 | 627 | |||
| BC132629.1 | - | 7 | 633 | 40130906 | ||

Tmprss11f
Genomic Map