siRNA Details
| Gene Symbol: Tmprss11e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172880.2 | NP_766468.1 | 8 | 776 | ||
| GenBank mRNA | AK037235.1 | - | 8 | 778 | ||
| AK162701.1 | - | 8 | 774 | |||
| AY787860.1 | - | 8 | 784 | |||
| BC115432.1 | - | 8 | 770 | 40094164 | ||
| BC115433.1 | - | 8 | 770 | 40094170 | ||

Tmprss11e
Genomic Map