siRNA Details
| Gene Symbol: Tmprss11e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172880.2 | NP_766468.1 | 2 | 108 | ||
| GenBank mRNA | AK037235.1 | - | 2 | 110 | ||
| AK162701.1 | - | 2 | 105 | |||
| AY787860.1 | - | 2 | 116 | |||
| BC115432.1 | - | 2 | 102 | 40094164 | ||
| BC115433.1 | - | 2 | 102 | 40094170 | ||

Tmprss11e
Genomic Map