siRNA Details
| Gene Symbol: Sbno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081203.1 | NP_001074672.1 | 25 | 3304 | ||
| GenBank mRNA | AB188811.1 | - | 24 | 3233 | ||
| AK133096.1 | - | 17 | 2128 | |||
| AK143120.1 | - | 7 | 617 | |||
| BC027253.1 | - | 7 | 692 | |||
| BC063073.1 | - | 4 | 242 | |||
| BC138418.1 | - | 25 | 3361 | 8861438 | ||

Sbno1
Genomic Map