siRNA Details
| Gene Symbol: Sbno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081203.1 | NP_001074672.1 | 10 | 1306 | ||
| GenBank mRNA | AB188811.1 | - | 9 | 1235 | ||
| AK077133.1 | - | 10 | 1312 | |||
| AK133096.1 | - | 2 | 129 | |||
| AK164542.1 | - | 9 | 2746 | |||
| AK165852.1 | - | 9 | 1245 | |||
| BC021875.1 | - | 9 | 1252 | 4212192 | ||
| BC023706.1 | - | 6 | 674 | |||
| BC106156.1 | - | 10 | 1365 | |||
| BC138418.1 | - | 10 | 1363 | 8861438 | ||

Sbno1
Genomic Map