siRNA Details
| Gene Symbol: A430033K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183025.2 | NP_898846.2 | 3, 4 | 348 | ||
| GenBank mRNA | AK039936.1 | - | 334 | |||
| BC055299.1 | - | 3, 4 | 313 | 6492709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A430033K04Rik| Gene Symbol: A430033K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183025.2 | NP_898846.2 | 3, 4 | 348 | ||
| GenBank mRNA | AK039936.1 | - | 334 | |||
| BC055299.1 | - | 3, 4 | 313 | 6492709 | ||