siRNA Details
Gene Symbol: Slc29a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146257.2 | NP_666369.1 | 7 | 841 | ||
GenBank mRNA | BC025599.1 | - | 7 | 785 | 5250967 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc29a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146257.2 | NP_666369.1 | 7 | 841 | ||
GenBank mRNA | BC025599.1 | - | 7 | 785 | 5250967 |