siRNA Details
| Gene Symbol: Ppm1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175523.4 | NP_780732.1 | 2 | 369 | ||
| GenBank mRNA | AK044610.1 | - | 2 | 439 | ||
| BC092238.1 | - | 2 | 369 | 30642218 | ||
| BC138473.1 | - | 1 | 175 | 8861493 | ||
| BC138474.1 | - | 1 | 175 | 8861494 | ||

Ppm1k
Genomic Map