siRNA Details
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172891.2 | NP_766479.1 | 8 | 1877 | ||
GenBank mRNA | AK033606.1 | - | 8 | 1877 | ||
AK035247.1 | - | 8 | 1038 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172891.2 | NP_766479.1 | 8 | 1877 | ||
GenBank mRNA | AK033606.1 | - | 8 | 1877 | ||
AK035247.1 | - | 8 | 1038 |