siRNA Details
Gene Symbol: Ppp1r9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181595.3 | NP_853626.1 | 2 | 1216 | ||
GenBank mRNA | AB091828.1 | - | 2 | 1216 | ||
AK030399.1 | - | 2 | 1205 | |||
AK135170.1 | - | 2 | 1116 | |||
AK138313.1 | - | 2 | 1145 | |||
AK158559.1 | - | 2 | 1164 | |||
AY308065.1 | - | 1 | 891 | |||
BC053748.1 | - | 2 | 1104 | 6331919 |
