siRNA Details
Gene Symbol: Plxna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175750.3 | NP_786926.2 | 17, 18 | 4226 | ||
GenBank mRNA | AB073228.1 | - | 17, 18 | 4225 | ||
AK051614.1 | - | 2, 3 | 129 | |||
BC141298.1 | - | 17, 18 | 3524 | 9055910 |
