siRNA Details
Gene Symbol: Plxna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175750.3 | NP_786926.2 | 2 | 1608 | ||
GenBank mRNA | AB073228.1 | - | 2 | 1607 | ||
BC141298.1 | - | 2 | 906 | 9055910 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plxna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175750.3 | NP_786926.2 | 2 | 1608 | ||
GenBank mRNA | AB073228.1 | - | 2 | 1607 | ||
BC141298.1 | - | 2 | 906 | 9055910 |