siRNA Details
| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172892.3 | NP_766480.1 | 8 | 1487 | ||
| GenBank mRNA | AK036354.1 | - | 8 | 1489 | ||
| AK038937.1 | - | 8 | 1492 | |||
| BC089161.1 | - | 8 | 1476 | 30440173 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc13a4| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172892.3 | NP_766480.1 | 8 | 1487 | ||
| GenBank mRNA | AK036354.1 | - | 8 | 1489 | ||
| AK038937.1 | - | 8 | 1492 | |||
| BC089161.1 | - | 8 | 1476 | 30440173 | ||