siRNA Details
Gene Symbol: Zfp324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178732.3 | NP_848847.3 | 4 | 1594 | ||
GenBank mRNA | AK044240.1 | - | 4 | 1591 | ||
AK051488.1 | - | 4 | 1617 | |||
AK052251.1 | - | 3 | 1487 | |||
AK054407.1 | - | 4 | 1639 | |||
AK085053.1 | - | 3 | 1490 | |||
AK152107.1 | - | 4 | 1620 | |||
AK152696.1 | - | 4 | 1537 | |||
AK165922.1 | - | 4 | 1590 | |||
BC053093.1 | - | 4 | 1594 | 6400158 |
