siRNA Details
Gene Symbol: Zfp536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172385.2 | NP_759017.1 | 6 | 4005 | ||
GenBank mRNA | AK030633.1 | - | 6 | 4006 | ||
AK034508.1 | - | 6 | 3949 | |||
AK042625.1 | - | 3 | 2065 | |||
AK042860.1 | - | 1 | 706 | |||
AK122270.1 | - | 7 | 4012 | |||
AK163177.1 | - | 1 | 472 | |||
BC033594.1 | - | 6 | 3927 | 5367892 |
