siRNA Details
| Gene Symbol: Mctp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024703.1 | NP_001019874.1 | 2 | 293 | ||
| GenBank mRNA | AK156983.1 | - | 2 | 307 | ||
| BC086658.1 | - | 2 | 293 | 30546156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mctp2| Gene Symbol: Mctp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024703.1 | NP_001019874.1 | 2 | 293 | ||
| GenBank mRNA | AK156983.1 | - | 2 | 307 | ||
| BC086658.1 | - | 2 | 293 | 30546156 | ||