siRNA Details
| Gene Symbol: Tsku | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024619.3 | NP_001019790.1 | 2 | 491 | ||
| NM_001168539.1 | NP_001162011.1 | 2 | 495 | |||
| NM_001168540.1 | NP_001162012.1 | 3 | 601 | |||
| NM_001168541.1 | NP_001162013.1 | 4 | 741 | |||
| GenBank mRNA | AB214184.1 | - | 1 | 436 | ||
| AK035461.1 | - | 2 | 492 | |||
| AK053973.1 | - | 4 | 742 | |||
| AK137208.1 | - | 2 | 492 | |||
| BC116310.1 | - | 2 | 503 | 40095310 | ||
| BC116311.1 | - | 3 | 613 | 40095311 | ||
| BC129977.1 | - | 2 | 503 | 40095312 | ||
| BC129978.1 | - | 3 | 613 | 40095313 | ||

Tsku
Genomic Map