siRNA Details
| Gene Symbol: Ubqlnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198624.3 | NP_941026.2 | 1 | 1084 | ||
| GenBank mRNA | AK076645.1 | - | 1 | 1075 | ||
| BC111901.1 | - | 1 | 1012 | 40062172 | ||
| BC113134.1 | - | 1 | 1015 | 40062178 | ||

Ubqlnl
Genomic Map