siRNA Details
| Gene Symbol: Myo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081397.1 | NP_001074866.1 | 25 | 3299 | ||
| GenBank mRNA | AB429291.1 | - | 22 | 2805 | ||
| AK034257.1 | - | 2 | 173 | |||
| AK082350.1 | - | 25 | 3035 | |||
| AK220356.1 | - | 6 | 731 | |||
| BC082311.1 | - | 25 | 3165 | |||
| BC151049.1 | - | 25 | 3253 | 9087962 | ||
| BC151051.1 | - | 25 | 3254 | 9087964 | ||
| BC157966.1 | - | 25 | 3254 | 9007290 | ||
| BC172122.1 | - | 25 | 3254 | 9054408 | ||

Myo16
Genomic Map