siRNA Details
| Gene Symbol: Mcph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173189.2 | NP_775281.2 | 8 | 1870 | ||
| GenBank mRNA | AK042026.1 | - | 7 | 1764 | ||
| AK083572.1 | - | 8 | 1913 | |||
| AK147997.1 | - | 8 | 1462 | |||
| AK148021.1 | - | 8 | 1468 | |||
| AY070216.1 | - | 8 | 1450 | |||
| BC056924.1 | - | 8 | 1870 | 5697978 | ||

Mcph1
Genomic Map