siRNA Details
| Gene Symbol: Erlin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153592.2 | NP_705820.1 | 6 | 519 | ||
| GenBank mRNA | AK030557.1 | - | 6 | 463 | ||
| BC036333.1 | - | 6 | 484 | 3590904 | ||
| BC038374.1 | - | 6 | 459 | 5355446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erlin2| Gene Symbol: Erlin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153592.2 | NP_705820.1 | 6 | 519 | ||
| GenBank mRNA | AK030557.1 | - | 6 | 463 | ||
| BC036333.1 | - | 6 | 484 | 3590904 | ||
| BC038374.1 | - | 6 | 459 | 5355446 | ||