siRNA Details
Gene Symbol: Triml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177742.4 | NP_808410.1 | 6 | 1240 | ||
GenBank mRNA | AK077092.1 | - | 6 | 1242 | ||
BC050188.1 | - | 4 | 1519 | |||
BC141202.1 | - | 6 | 1219 | 9055814 | ||
BC145531.1 | - | 6 | 1222 | 9054094 |
