siRNA Details
| Gene Symbol: Triml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177742.4 | NP_808410.1 | 6 | 1480 | ||
| GenBank mRNA | AK077092.1 | - | 6 | 1482 | ||
| BC050188.1 | - | 4 | 1759 | |||
| BC141202.1 | - | 6 | 1459 | 9055814 | ||
| BC145531.1 | - | 6 | 1462 | 9054094 | ||

Triml1
Genomic Map