siRNA Details
| Gene Symbol: Triml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177742.4 | NP_808410.1 | 1 | 276 | ||
| GenBank mRNA | AK077092.1 | - | 1 | 278 | ||
| BC141202.1 | - | 1 | 255 | 9055814 | ||
| BC145531.1 | - | 1 | 255 | 9054094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Triml1| Gene Symbol: Triml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177742.4 | NP_808410.1 | 1 | 276 | ||
| GenBank mRNA | AK077092.1 | - | 1 | 278 | ||
| BC141202.1 | - | 1 | 255 | 9055814 | ||
| BC145531.1 | - | 1 | 255 | 9054094 | ||