siRNA Details
| Gene Symbol: Wdr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172152.1 | NP_001165623.1 | 12 | 1719 | ||
| NM_028220.3 | NP_082496.3 | 13 | 1742 | |||
| GenBank mRNA | AK044241.1 | - | 12 | 1720 | ||
| BC028268.1 | - | 13 | 1753 | 5366098 | ||

Wdr17
Genomic Map