siRNA Details
Gene Symbol: Adam29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175939.3 | NP_787953.2 | 2 | 1680 | ||
GenBank mRNA | AY190759.1 | - | 2 | 1664 | ||
BC119089.1 | - | 2 | 1661 | 8733838 | ||
BC119091.1 | - | 2 | 1661 | 8733840 |
