siRNA Details
| Gene Symbol: Pkd1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039700.2 | NP_001034789.2 | 16, 17 | 3617 | ||
| NM_001286454.1 | NP_001273383.1 | 15, 16 | 3587 | |||
| NM_181544.2 | NP_853522.2 | 15, 16 | 3587 | |||
| GenBank mRNA | AB290926.1 | - | 15, 16 | 3587 | ||
| AY164486.1 | - | 15, 16 | 3587 | |||
| DQ382344.1 | - | 15, 16 | 3587 | |||
| DQ382345.1 | - | 16, 17 | 3617 | |||
| DQ382346.1 | - | 16, 17 | 3599 | |||
| DQ382347.1 | - | 7, 8 | 1133 | |||

Pkd1l3
Genomic Map