siRNA Details
| Gene Symbol: Phlpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122594.2 | NP_001116066.2 | 5 | 813 | ||
| GenBank mRNA | AK048260.1 | - | 5 | 770 | ||
| AK048472.1 | - | 5 | 814 | |||
| BC117961.1 | - | 5 | 813 | 40098281 | ||
| BC117962.1 | - | 5 | 805 | 40098282 | ||

Phlpp2
Genomic Map