siRNA Details
| Gene Symbol: Bsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178245.3 | NP_839976.1 | 1 | 198 | ||
| GenBank mRNA | AY233396.1 | - | 1 | 198 | ||
| BC104386.1 | - | 1 | 196 | 40050191 | ||
| BC104387.1 | - | 1 | 196 | 40050192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bsx| Gene Symbol: Bsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178245.3 | NP_839976.1 | 1 | 198 | ||
| GenBank mRNA | AY233396.1 | - | 1 | 198 | ||
| BC104386.1 | - | 1 | 196 | 40050191 | ||
| BC104387.1 | - | 1 | 196 | 40050192 | ||