siRNA Details
| Gene Symbol: Npat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081152.1 | NP_001074621.1 | 4, 5 | 406 | ||
| GenBank mRNA | AK033012.1 | - | 4, 5 | 386 | ||
| AK080294.1 | - | 4, 5 | 407 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npat| Gene Symbol: Npat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081152.1 | NP_001074621.1 | 4, 5 | 406 | ||
| GenBank mRNA | AK033012.1 | - | 4, 5 | 386 | ||
| AK080294.1 | - | 4, 5 | 407 | |||