siRNA Details
Gene Symbol: Npat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081152.1 | NP_001074621.1 | 8 | 817 | ||
GenBank mRNA | AK033012.1 | - | 8 | 797 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Npat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081152.1 | NP_001074621.1 | 8 | 817 | ||
GenBank mRNA | AK033012.1 | - | 8 | 797 |