siRNA Details
| Gene Symbol: AI118078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172923.3 | NP_766511.1 | 11 | 1261 | ||
| GenBank mRNA | AK036040.1 | - | 11 | 1261 | ||
| BC068128.1 | - | 11 | 3270 | 6835202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI118078| Gene Symbol: AI118078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172923.3 | NP_766511.1 | 11 | 1261 | ||
| GenBank mRNA | AK036040.1 | - | 11 | 1261 | ||
| BC068128.1 | - | 11 | 3270 | 6835202 | ||