siRNA Details
| Gene Symbol: Col6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102607.1 | NP_001096077.1 | 4 | 950 | ||
| NM_172927.3 | NP_766515.2 | 4 | 985 | |||
| GenBank mRNA | AK054356.1 | - | 4 | 986 | ||
| AK087828.1 | - | 4 | 1026 | |||
| AM748260.1 | - | 2 | 670 | |||
| BC150695.1 | - | 3 | 769 | 9087606 | ||

Col6a6
Genomic Map