siRNA Details
| Gene Symbol: Fam198a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177743.5 | NP_808411.1 | 5 | 1681 | ||
| GenBank mRNA | AK050223.1 | - | 5 | 1669 | ||
| AK134876.1 | - | 5 | 1736 | |||
| AK158406.1 | - | 5 | 1683 | |||
| BC113767.1 | - | 4 | 1530 | 40058404 | ||

Fam198a
Genomic Map