siRNA Details
Gene Symbol: Zfp300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183185.3 | NP_899008.2 | 5 | 2167 | ||
GenBank mRNA | AK053018.1 | - | 5 | 2168 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183185.3 | NP_899008.2 | 5 | 2167 | ||
GenBank mRNA | AK053018.1 | - | 5 | 2168 |