siRNA Details
| Gene Symbol: Gpr101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033360.3 | NP_001028532.1 | 2 | 870 | ||
| GenBank mRNA | AK138185.1 | - | 2 | 814 | ||
| AK138370.1 | - | 2 | 872 | |||
| AY255576.1 | - | 1 | 161 | |||
| BC139013.1 | - | 1 | 662 | 8862035 | ||
| BC139014.1 | - | 1 | 662 | 8862036 | ||
| DQ198079.1 | - | 2 | 830 | |||
More Information
Set Membership: |
|

Gpr101
Genomic Map