siRNA Details
| Gene Symbol: Zc3h12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034907.2 | NP_001030079.2 | 8 | 1882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h12b| Gene Symbol: Zc3h12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034907.2 | NP_001030079.2 | 8 | 1882 | ||