siRNA Details
| Gene Symbol: Hdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080549.2 | NP_001074018.1 | 3 | 1078 | ||
| NM_001290459.1 | NP_001277388.1 | 2 | 931 | |||
| GenBank mRNA | AK083436.1 | - | 2 | 919 | ||
| BC116300.1 | - | 3 | 1021 | 40095063 | ||
| BC116301.1 | - | 2 | 874 | 40095064 | ||

Hdx
Genomic Map